missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ MICROGARD™ 1500 Coveralls, Model 138

Product Code. 15661784 Shop All Ansell Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15661784 White Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15661784 Supplier Ansell™ Supplier No. WH15S0013809
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

SMS fabric proven to filter 100% of particles larger than 3 microns, while air and water vapor permeable and optimized for body fit, AlphaTec™ MICROGARD™ 1500 Coveralls are designed for workers involved in handling asbestos, general maintenance, construction, and contract cleaning.

  • Three-piece hood
  • Elasticated hood, wrists, waist, and ankles
  • Two-way front zipper with resealable storm flap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class Category III
Resistance Abrasion, Acid
Certifications/Compliance EN ISO 13982-1: 2004+A1:2010, Type 5; EN 13034: 2005+A1: 2009, Type 6
Description Alphatec™ 1500 coveralls have been designedfor workers involved in the stripping, clear up orhandling of asbestos, general maintenance,construction and contract cleaning.
Latex Free Yes
Model Model 138
No. of Pockets None
Size (English) Small
Sleeve Style Long sleeves
For Use With (Application) Asbestos Handling, General Maintenance, Construction, and Contract Cleaning
Weight (English) 0.006 oz.
Weight (Metric) 0.169 g
Quantity 50 Cs.
Style Alphatec™ 1500
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.