missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™MICROGARD™2500 Standard Model 111 Suit, White

Product Code. p-7146547
Click to view available options
Size:
X-Small
Small
Unit Size:
Pack of 35
This item is not returnable. View return policy

Product Code. 15886474

Brand: Ansell™ WH25T0011101

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

MICROGARD™ 2500 is a unique material offeringexceptional mechanical strength, liquid andparticulate protection

  • 3-piece hood
  • Elasticated hood, wrist, waist and ankles
  • 2-way front zip with resealable storm flap
  • Finger loops
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical-resistant Coveralls
Size X-Small
Gender Unisex
Material Polypropylene Laminate
Color White
Ankle Style Elastic
Cuff Style Finger Loops, Elastic
Hood Type 3-panel
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class Category III
Resistance Bloodborne Pathogens, Anti-static, Liquid, Chemicals, Acid
Collar Type Hooded
Certifications/Compliance EN1073-2; EN1149-5; EN14126; Type 4-B; Type 5-B; Type 6-B
Description Suit with 3 piece, elasticated hood, elasticated wrist, waist and ankles. Features finger loops.
Latex Free Yes
No. of Pockets None
Size (English) X-Small
Sleeve Style Long sleeves
For Use With (Application) Viral contaminated areas (including Avian influenza), biological protection, nuclear industry, medical research
Weight (Metric) 0.304 g
Quantity 35/Cs.
Style WH25T-00111-01
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.