Learn More
Ansell AlphaTec™ Sure Step Overshoes - Model 403
Product Code. 17161025
Change view
Click to view available options
Size:
46 to 48
Unit Size:
250 pairs
Product Code. Size Unit Size
17161025 46 to 48 250 pairs
1 options
Product Code. 17161025 Supplier Ansell Supplier No. WHSSS0040305
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 19-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AlphaTec™ Sure Step Overshoes are disposable shoes that provides slip-resistance to the underside of the feet.
  • A disposable shoe that provides slip-resistance to the underside of the feet.
TRUSTED_SUSTAINABILITY
Specifications
Size 46 to 48
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Certifications/Compliance (Europe) Not certified PPE
Resistance Chemical-resistant
Product Type Controlled Environments Shoe Covers