Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
Product Code. 17730768
Change view
Click to view available options
Description:
384-well PCR Plate Carrier
96-Well PCR Plate Carrier
Standard Microplate Carrier
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17730768 Standard Microplate Carrier Each
11992281 96-Well PCR Plate Carrier Each
11942291 384-well PCR Plate Carrier Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17730768 Supplier Thermo Scientific™ Supplier No. AB0827
  • ONLINE EXCLUSIVE PRICE
    415.00 €
    379.99 € / Pack of 1
    Save 35.01 €
    8% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 year
TRUSTED_SUSTAINABILITY
Specifications
Description Standard Microplate Carrier
For Use With (Equipment) ALPS™ Heat Sealer
Product Type Carrier