missing translation for 'onlineSavingsMsg'
Learn More

Altanserin hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18015762
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18015762

Brand: Tocris Bioscience 1809/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-HT2A receptor antagonist

Potent and selective 5-HT2A receptor antagonist (Ki values are 0.13, 4.55, 40, 62 and 1570 nM at 5-HT2A, α1, 5-HT2C, D2 and 5-HT1A respectively). Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135280-78-2
Molecular Formula C22H23ClFN3O2S
Molecular Weight (g/mol) 447.953
InChI Key JFPPLMAMMZZOEA-UHFFFAOYSA-N
Synonym altanserin hydrochloride, unii-xul9717f5k, dsstox_cid_22592, dsstox_rid_80054, dsstox_gsid_42592, 4 1h-quinazolinone, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-, hydrochloride 1:1, 3-2-4-4-fluorobenzoyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorophenyl carbonyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone hydrochloride
PubChem CID 24978536
IUPAC Name 3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-2-sulfanylidene-1H-quinazolin-4-one;hydrochloride
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=S.Cl

Specifications

Quantity 50 mg
Formula Weight 447.95
Percent Purity >98%
Chemical Name or Material Altanserin hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.