missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Altanserin hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective 5-HT2A receptor antagonist (Ki values are 0.13, 4.55, 40, 62 and 1570 nM at 5-HT2A, α1, 5-HT2C, D2 and 5-HT1A respectively). Centrally active following systemic administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 447.95 |
| Percent Purity | >98% |
| Chemical Name or Material | Altanserin hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction