missing translation for 'onlineSavingsMsg'
Learn More

(-)-Altenuene, TRC

Product Code. 18061010
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
0.25mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18061010 1 mg 1mg
18050200 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18061010 Supplier Toronto Research Chemicals Supplier No. TRCA5757401MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 889101-41-1
Molecular Formula C15H16O6
Molecular Weight (g/mol) 292.28
Synonym 6H-Dibenzo[b,d]pyran-6-one, 2,3,4,4a-tetrahydro-2,3,7-trihydroxy-9-methoxy-4a-methyl-, (2R,3R,4aR)-, (2R,3R,4aR)-2,3,4,4a-Tetrahydro-2,3,7-trihydroxy-9-methoxy-4a-methyl-6H-dibenzo[b,d]pyran-6-one, (-)-Altenuene
SMILES COc1cc(O)c2C(=O)O[C@]3(C)C[C@@H](O)[C@H](O)C=C3c2c1

Specifications

Quantity 1 mg
Percent Purity >95%
Chemical Name or Material (-)-Altenuene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.