Specifications
| Quantity | 25 Pk. |
| Type | Aluminum Sheet |
| For Use With (Application) | Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates |
| Specific Surface | 15,000 cm2/g |
| Dimensions (L x W) | 20 x 20 cm |