missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acetate, 90%, basic

Product Code. 10765832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10765832 250 g Glass bottle 250g
10667020 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10765832 Supplier Thermo Scientific Acros Supplier No. 428192500
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-03-0
Molecular Formula C4H7AlO5
Molecular Weight (g/mol) 162.08
MDL Number MFCD00008688
InChI Key HQQUTGFAWJNQIP-UHFFFAOYSA-K
Synonym aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate
PubChem CID 18502856
IUPAC Name (acetyloxy)(hydroxy)alumanyl acetate
SMILES CC(=O)O[Al](O)OC(C)=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Quantity 250 g
Packaging Glass bottle
Assay Percent Range 90%
Merck Index 15,359
Solubility Information Solubility in water: slightly soluble
Formula Weight 162.08
Percent Purity 90%
Chemical Name or Material Aluminum acetate, Basic
compliance-icons
Product Identifier
  • Aluminium acetate, basic
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.