missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acetylacetonate, 97%

Product Code. 10348430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348430 5 g Glass bottle 5g
10781661 100 g Plastic bottle 100g
10209203 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10348430 Supplier Thermo Scientific Acros Supplier No. 196970050
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym aluminum 2, 4-pentanedionate
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color White to Cream
Boiling Point 315.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Glass bottle
Linear Formula (CH3COCH2COCH2)3Al
Quantity 5 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: 4.26 g/100g alcohol (20°C), 36.45 g/100g benzene (20°C), insoluble in petroleum ether
Formula Weight 324.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Aluminum acetylacetonate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium acetylacetonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.