missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Ammonium Sulfate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10131610 Shop All Fisher Chemical Products
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
500 g
Pakningsstørrelse:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
10131610 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10131610 Leverandør Fisher Chemical Leverandørnr. A/1880/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

H4AlNO8S2.12H2O, CAS Number-7784-26-1, 500g, 200 deg.C, White, 453.33g/mol, 93.5 deg.C, 149958, Plastic powder jar, 4.6, Solid

TRUSTED_SUSTAINABILITY

Tekniske data

Boiling Point 200°C
Melting Point 93.5°C
CAS 7784-26-1
Color White
pH 4.6
Physical Form Solid
Quantity 500 g
Molecular Formula AlH28NO20S2
MDL Number 149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name aluminium(3+) ammonium dodecahydrate disulfate
Molecular Weight (g/mol) 453.31
Formula Weight 453.33g/mol
Packaging Plastic powder jar
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Aluminium ammonium sulfate dodecahydrate
Signalord
  • Warning
Farekategori
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.