missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ammonium sulfate dodecahydrate, 99%, pure

Product Code. 10628704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10628704 100 g Plastic bottle 100g
10134500 5 g Glass Bottle 5g
10450481 1 kg Plastic bottle 1kg
10460481 2.5 kg Plastic bottle 2.5kg
11934261 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10628704 Supplier Thermo Scientific Acros Supplier No. 206261000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7784-26-1 | H4AlNO8S2·12H2O | 453.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-26-1
MDL Number MFCD00149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

Specifications

Boiling Point 200.0°C
Melting Point 93.5°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.5% min. (Complexometry)
Molecular Formula H4AlNO8S2·12H2O
Linear Formula AlNH4(SO4)2·12H2O
Merck Index 15,323
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol,practically insoluble in alcohol
Molecular Weight (g/mol) 453.33
Formula Weight 453.33
Percent Purity 99%
Grade Pure
Loss on Drying 45% to 48% (2 g, 300°C)
Packaging Plastic bottle
Specific Gravity 1.64
Density 1.6400g/mL
Chemical Name or Material Aluminum ammonium sulfate dodecahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium ammonium sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Aluminium ammonium sulfate dodecahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.