missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, 98+%, Pure, Anhydrous, Fisher Chemical

Product Code. 10558030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10558030 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10558030 Supplier Fisher Chemical Supplier No. A/2000/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 194°C
CAS 7446-70-0
Color Yellow
pH 2.4
Physical Form Solid
Quantity 500 g
Molecular Formula AlCl3
MDL Number 3422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES [Al+3].[Cl-].[Cl-].[Cl-]
IUPAC Name aluminium(3+) trichloride
Molecular Weight (g/mol) 133.33
Formula Weight 133.34g/mol
Packaging Amber glass bottle
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium chloride
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.