Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Quantity | 1 kg |
| Packaging | Glass bottle |
| Specific Gravity | 2.39 |
| Assay Percent Range | 98.5% min. (Argentometry) |
| Density | 2.3900g/mL |
| Linear Formula | AlCl3·6H2O |
| Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 |
| Merck Index | 15,333 |
| Solubility Information | Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene |
| IUPAC Name | aluminium(3+) hexahydrate trichloride |
| Afficher plus |
Sécurité et traitement
- Aluminium chloride hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P264-Wash face, hands and any thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.