missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Quantity | 500 g |
| Packaging | Glass bottle |
| Specific Gravity | 2.39 |
| Assay Percent Range | 98.5% min. (Argentometry) |
| Density | 2.3900g/mL |
| Linear Formula | AlCl3·6H2O |
| Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 |
| Merck Index | 15,333 |
| Solubility Information | Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene |
| IUPAC Name | aluminium(3+) hexahydrate trichloride |
| Show More |
Safety and Handling
Product Identifier
- Aluminium chloride hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P264-Wash face, hands and any thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?