missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, technical, foil, ≈0.1mm thick, SLR, Fisher Chemical™

Product Code. 10214252 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10214252 250 g 250g
10030000 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10214252 Supplier Fisher Chemical Supplier No. A/1570/50
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Gray-Silver
Melting Point 660°C
Boiling Point 2327°C
Quantity 250 g
Packaging Plastic container
Form Solid
Formula Weight 26.97g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.