missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
Specifications
Specifications
| Melting Point | 129.0°C to 136.0°C |
| Color | White |
| Boiling Point | 125.0°C to 130.0°C (38.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.99% |
| Packaging | Glass bottle |
| Linear Formula | [(CH3)2CHO]3Al |
| Quantity | 50 g |
| Fieser | 01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16 |
| Merck Index | 15,342 |
| Show More |
Safety and Handling
Product Identifier
- Aluminium isopropoxide
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: 2-Propanol, aluminumsalt
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?