missing translation for 'onlineSavingsMsg'
Learn More

Aluminum isopropoxide, 99.99%, (trace metal basis)

Product Code. 10716433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10716433 250 g Glass Bottle 250g
10195422 10 g Glass Bottle 10g
10546413 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10716433 Supplier Thermo Scientific Acros Supplier No. 212232500
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 555-31-7 | C9H21AlO3 | 204.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-31-7
Molecular Formula C9H21AlO3
Molecular Weight (g/mol) 204.25
MDL Number MFCD00008870
InChI Key SMZOGRDCAXLAAR-UHFFFAOYSA-N
Synonym aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
PubChem CID 11143
IUPAC Name aluminum;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Specifications

Melting Point 129.0°C to 136.0°C
Color White
Boiling Point 125.0°C to 130.0°C (38.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99.99%
Packaging Glass Bottle
Linear Formula [(CH3)2CHO]3Al
Quantity 250 g
Fieser 01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16
Merck Index 15,342
Solubility Information Solubility in water: reacts. Other solubilities: freely soluble in ethanol
Formula Weight 204.25
Percent Purity 99.99%
Grade Trace Metal Basis
Physical Form Crystalline Powder
Chemical Name or Material Aluminum isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.