missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitrate Nonahydrate, 98+%, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072791 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
5kg
This item is not returnable. View return policy

Product Code. 11403663

Brand: Fisher Chemical A/2200/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.13
MDL Number 149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
IUPAC Name aluminium(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 73°C
pH 2.5 to 3.5
Physical Form Solid
Quantity 250 g
Formula Weight 375.13g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Aluminium nitrate nonahydrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aluminium nitrate nonahydrate, Aluminumnitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.