missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent

Product Code. 10647411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647411 500 g Plastic bottle 500g
10301601 5 g Glass Bottle 5g
10647454 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10647411 Supplier Thermo Scientific Acros Supplier No. 423265000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7784-24-9 | AlKO8S2·12H2O | 474.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlKO8S2·12H2O
Molecular Weight (g/mol) 474.39
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92°C
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula AlK(SO4)2·12H2O
Merck Index 15, 354
Solubility Information Solubility in water: 140g/L (20°C). Other solubilities: freely soluble in glycerol,insoluble in alcohol
Formula Weight 474.39
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Aluminum potassium sulfate dodecahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.