missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 11443713 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
Unit Size:
1kg
3kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443713 500 g 500g
10765321 1 kg 1kg
10588410 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11443713 Supplier Fisher Chemical Supplier No. A/2400/53
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Aluminum Potassium Sulfate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
MDL Number 149143

Specifications

Melting Point 92°C
Color White
pH 3.0 to 3.5
Physical Form Solid
Quantity 500 g
Molecular Formula AlKO8S2·12H2O
Formula Weight 474.39g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.