missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate, pure, anhydrous

Product Code. 15572353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15572353 1 kg 1kg
15582353 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15572353 Supplier Thermo Scientific Acros Supplier No. 458990010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10043-67-1 | AlKO8S2 | 258.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-67-1
Molecular Formula AlKO8S2
Molecular Weight (g/mol) 258.21
InChI Key GRLPQNLYRHEGIJ-UHFFFAOYSA-J
Synonym potassium alum, potash alum, aluminum potassium sulfate, potassium aluminum sulfate, burnt potassium alum, aluminum potassium disulfate, exsiccated alum, alum potassium, burnt alum, aluminum potassium alum
PubChem CID 24856
ChEBI CHEBI:86463
IUPAC Name aluminum;potassium;disulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Boiling Point 200.0°C
Melting Point 92.0°C
Quantity 1 kg
Solubility Information Solubility in water: 50g/L (20°C).
Formula Weight 258.21
Percent Purity ≥90% (Complexometry)
Grade Pure
Loss on Drying 1% max.
Chemical Name or Material Aluminum potassium sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.