missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Powder, Coarse, Grease Free, Technical, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Specifications
Specifications
| Color | Gray to Silver |
| Melting Point | 660°C |
| Boiling Point | 2467°C |
| Quantity | 250 g |
| Packaging | Glass Jar |
| Form | Solid |
| Formula Weight | 26.98g/mol |
Safety and Handling
Product Identifier
- Aluminium metal, fine powder
- Danger
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P240-Ground/bond container and receiving equipment.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction