missing translation for 'onlineSavingsMsg'
Learn More

Aluminum trifluoromethanesulfonate, 99%

Product Code. 10404905 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10404905 10 g Plastic bottle 10g
10005643 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10404905 Supplier Thermo Scientific Acros Supplier No. 383260100
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74974-61-1 | C3AlF9O9S3 | 474.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74974-61-1
Molecular Formula C3AlF9O9S3
Molecular Weight (g/mol) 474.19
MDL Number MFCD00143596
InChI Key FKOASGGZYSYPBI-UHFFFAOYSA-K
Synonym aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate
PubChem CID 2737634
IUPAC Name aluminum;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point 300.0°C
Color White
Physical Form Powder
Quantity 10 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98.5% min. (Complexometry)
Solubility Information Solubility in water: insoluble
Formula Weight 474.19
Percent Purity 99%
Chemical Name or Material Aluminum trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.