missing translation for 'onlineSavingsMsg'
Learn More

Aluminon

Product Code. 10090610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10090610 25 g Glass bottle 25g
10548090 100 g Glass bottle 100g
10530994 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10090610 Supplier Thermo Scientific Acros Supplier No. 103060250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.438
MDL Number MFCD00040925
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym Aurintricarboxylic acid, triammonium salt
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220.0°C to 225.0°C
Beilstein 10, IV, 4161
Merck Index 15, 318
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol, insoluble in petroleum ether
Infrared Spectrum Authentic
Formula Weight 473.44
Packaging Glass bottle
Suitability suitable for determination of aluminum
Color Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Aluminon
Show More Show Less
What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.

compliance-icons
Product Identifier
  • Aluminon
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.