missing translation for 'onlineSavingsMsg'
Learn More

Aluminon

Product Code. 11478366
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478366 250 g 250g
11458366 10 g 10g
11468366 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478366 Supplier Thermo Scientific Alfa Aesar Supplier No. A15401.30
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol

It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types.

Solubility
Soluble in water(800g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.438
MDL Number MFCD00040925
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym Aurintricarboxylic acid triammonium salt
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220°C to 225°C (decomposition)
Odor Odorless
Merck Index 14,322
Solubility Information Soluble in water(800g/L).
pH 5.3
Quantity 250 g
Formula Weight 473.44
Chemical Name or Material Aluminon
What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.