missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Acetate, Basic, Honeywell™ Fluka™

Product Code. 15672940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15672940 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672940 Supplier Honeywell-Fluka Supplier No. 289825100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 142-03-0
Quantity 100 g
Molecular Formula C4H7AlO5
Linear Formula Al(OH)(C2H3O2)2
MDL Number MFCD00008688
UN Number NONH for all modes of transport
Synonym aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate
InChI Key HQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILES CC(=O)O[Al](O)OC(C)=O
IUPAC Name (acetyloxy)(hydroxy)alumanyl acetate
Molecular Weight (g/mol) 162.08
PubChem CID 18502856
Formula Weight 162.08g/mol
Packaging Poly Bottle
Chemical Name or Material Aluminum Acetate, Basic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.