missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Si, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Aluminum, Al/Si, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Lead/Chromium/Tin/Titanium/Manganese/Zinc/Iron/Nickel/Copper/Magnesium/Silicon |
| CAS | 7429-90-5 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.004 %/Lead 0.083 %/Chromium 0.103 %/Tin 0.145 %/Titanium 0.166 %/Manganese 0.248 %/Zinc 0.53 %/Iron 0.767 %/Nickel 1.15 %/Copper 1.29 %/Magnesium 1.37 %/Silicon 10.42 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction