missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ammonium sulfate dodecahydrate, Reagent Grade

Product Code. 11361349
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361349 100 g 100g
11351349 500 g 500g
11341349 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361349 Supplier Thermo Scientific Alfa Aesar Supplier No. 013802.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-26-1 | AlH28NO20S2 | 453.313 g/mol

Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper platingAluminum ammonium sulfate dodecahydrate acts as a crystalline double sulfate. It is used in water purification, in vegetable glues, in porcelain cements, in deodorant, in tanning and in fireproofing textiles. It serves as an ingredient in animal repellant sprays. Further, it is used in the preparation of Alum (Al(OH)3) gel. In addition, it is used in cosmetic formulation, fertilizers, food preservation and in the heating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-26-1
Molecular Formula AlH28NO20S2
Molecular Weight (g/mol) 453.313
MDL Number MFCD00051042
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point 93°C to 94°C
Color White
Physical Form Crystals
Quantity 100 g
Solubility Information Soluble in water and glycerol. Insoluble in alcohol.
Formula Weight 453.33 (237.14 Anhydrous)
Grade Reagent
Density 1.65
Chemical Name or Material Aluminum ammonium sulfate dodecahydrate
compliance-icons
Product Identifier
  • Aluminum ammonium sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.