missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 954, C95400, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 954, C95400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Selenium/Arsenic/Cadmium/Zirconium/Beryllium/Bismuth/Silver/Magnesium/Antimony/Sulfur/Cobalt/Phosphorus/Tin/Lead/Chromium/Silicon/Zinc/Nickel/Manganese/Iron/Aluminium/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon -50 ppm/Selenium -30 ppm/Arsenic -20 ppm/Cadmium -8 ppm/Zirconium -7 ppm/Beryllium -4 ppm/Bismuth 0.0009 %/Silver 0.0012 %/Magnesium 0.0013 %/Antimony 0.0019 %/Sulfur 0.0019 %/Cobalt 0.002 %/Phosphorus 0.0031 %/Tin 0.0053 %/Lead 0.012 %/Chromium 0.022 %/Silicon 0.029 %/Zinc 0.029 %/Nickel 0.105 %/Manganese 0.393 %/Iron 3.66 %/Aluminium 10.55 %/Copper 85.1 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction