missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Aluminum Burette Holder

Product Code. 10314271
Change view
Click to view available options
Diameter (Metric):
0 to 13 mm
Material:
Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Material Unit Size
10314271 0 to 13 mm Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314271 Supplier Bochem™ Supplier No. 5600
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from aluminum. BOCHEM™ Aluminum Burette Holder assists in the dispensing of measured amounts of chemical solution by holding the burette in place.

Aluminum burette holder.

  • Aluminum
  • Holds two burettes
  • With bosshead
    • With PVC rolls
    • Distance between rolls is 170mm

Specifications

Quantity 1 Ea.
Material Aluminum
Diameter (Metric) 0 to 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.