missing translation for 'onlineSavingsMsg'
Learn More

Aluminum carbide, 99+% (metals basis)

Product Code. 11391569
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11391569

Brand: Thermo Scientific Alfa Aesar 014038.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1299-86-1 | C3Al4 | 143.96 g/mol

Aluminum carbide is used as an abrasive material for cutting tools. It is also used to reduce metal oxides and to prepare aluminum nitride and methane. It and also finds use in metallurgy. Further, it acts as a catalyst and a drying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1299-86-1
Molecular Formula C3Al4
Molecular Weight (g/mol) 143.96
MDL Number MFCD00003425
InChI Key CAVCGVPGBKGDTG-UHFFFAOYSA-N
Synonym aluminum carbide, 1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne, aluminum carbide,al4c3, al4c3 aluminum carbide, aluminum carbide, al4c3, aluminum carbide, powder, ∼325 mesh
PubChem CID 16685054
IUPAC Name alumanylidynemethyl(alumanylidynemethylalumanylidenemethylidene)alumane
SMILES [Al]#C[Al]=C=[Al]C#[Al]

Specifications

Melting Point 2100°C (>2200° (decomposition))
Color Gray
Quantity 50 g
UN Number UN1394
Merck Index 14,335
Solubility Information Soluble in dilute hydrochloric acid.
Formula Weight 143.96
Percent Purity ≥99%
Sensitivity Moisture sensitive
Density 2.36 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum carbide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminum carbide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.