missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride hexahydrate, Reagent Grade

Product Code. 11351649
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351649 500 g 500g
11341649 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351649 Supplier Thermo Scientific Alfa Aesar Supplier No. 012297.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 100°C (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Solubility Information Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43 (133.34 Anhydrous)
Grade Reagent
Odor Odorless
Density 2.398
Chemical Name or Material Aluminum chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.