Learn More
VACUUBRAND™ Aluminum Clamping Rings
Product Code. 10518381
Change view
Click to view available options
Size:
DN 10/16 mm
DN 20/25 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10518381 DN 20/25 mm Each
10419122 DN 10/16 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10518381 Supplier VACUUBRAND™ Supplier No. 20660001
  • ONLINE EXCLUSIVE PRICE
    25.85 €
    24.60 € / Pack of 1
    Save 1.25 €
    4% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A rapid connection that can be mounted without any tools for soft seals and indium seals.
  • Provided with wing nut
  • Available in two different sizes
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 20/25 mm
For Use With For Small KF Flange
Material Aluminum
Quantity 1 Each
Size DN 20/25 mm
Type Clamping Ring