missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Aluminum Crimp Seals

Product Code. 11318133
Change view
Click to view available options
Diameter (Metric) Outer:
13 mm
20 mm
Volume (Metric):
2 mL
5, 10, 20 mL
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Volume (Metric) unitSize
11318133 20 mm 5, 10, 20 mL Pack of 500
10294854 13 mm 2 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11318133 Supplier Azpack™ Supplier No. 11318133
Add to basket
Edge
Add to basket

Seals freeze drying and injection vials. Azpack™Aluminum Crimp Seals prevent leakage of hazardous and biological products.

  • For use with clear neutral glass vials
  • Aluminum
  • 500/Pack

Specifications

Color Silver
Diameter (Metric) Outer 20 mm
Material Aluminum
Volume (Metric) 5, 10, 20 mL
Closure Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.