missing translation for 'onlineSavingsMsg'
Learn More

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min

Product Code. p-7080482
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413760 100 g 100g
11403760 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413760 Supplier Thermo Scientific Alfa Aesar Supplier No. 089350.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14782-75-3 | C12H23AlO5

Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14782-75-3
Molecular Formula C12H23AlO5
MDL Number MFCD00135521

Specifications

Density 1.05 g/mL
Flash Point 50°C (122°F)
Odor Alcohol-like
Quantity 100 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 274.3
Percent Purity ≥9.6%
Physical Form Liquid
Chemical Name or Material Aluminum di (isopropoxide)acetoacetic ester chelate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.