missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)

Product Code. 11498450
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498450 100 x 500 mm Each
11480700 50 x 50 mm Each
11488450 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498450 Supplier Thermo Scientific Alfa Aesar Supplier No. 044332.HP
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 100 x 500 mm
Odor Odorless
Weight ≈3.4g/50x50mm
Form Foil
Percent Purity 99.9999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum foil, 0.5mm (0.02 in.) thick, Annealed, Temper: soft

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.