missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Foil, 0.81mm (0.032 in.) thick, , Thermo Scientific™

Product Code. 15436785
Change view
Click to view available options
Quantity:
300 x 300 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436785 300 x 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436785 Supplier Thermo Scientific Alfa Aesar Supplier No. 045149.HB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Form: Solid in various forms

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 11145-27-0
Appearance Silver gray foil
Density 2.7g/cm3 at 20°C
Quantity 300 x 300 mm
Odor Odorless
Form Foil
Molecular Formula AlSi
MDL Number MFCD00134029
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
Solubility Information Insoluble in water.
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
SMILES [Al].[Si]
IUPAC Name aluminum;silicon
Molecular Weight (g/mol) 55.067
PubChem CID 9793685
Chemical Name or Material Aluminum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.