missing translation for 'onlineSavingsMsg'
Learn More

Aluminum iodide, 99.99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11389399
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389399 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11389399 Supplier Thermo Scientific Alfa Aesar Supplier No. 089185.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Moisture and Light Sensitive | CAS: 7784-23-8 | AlI3 | 407.695 g/mol

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-23-8
Molecular Formula AlI3
Molecular Weight (g/mol) 407.695
MDL Number MFCD00003427
InChI Key CECABOMBVQNBEC-UHFFFAOYSA-K
Synonym aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
PubChem CID 82222
IUPAC Name triiodoalumane
SMILES [Al](I)(I)I

Specifications

Boiling Point 360°C
Melting Point 191°C
Color Brown-Purple to White
Physical Form Lump
Quantity 5 g
Odor Pungent
Sensitivity Light and moisture sensitive
Assay Percent Range >99.99% (Metals basis)
Density 3.98
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 407.69
Chemical Name or Material Aluminum iodide
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aluminum iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.