missing translation for 'onlineSavingsMsg'
Learn More

Aluminum isopropoxide, 99.99+% (metals basis)

Product Code. 11332598
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332598 25 g 25g
11322598 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332598 Supplier Thermo Scientific Alfa Aesar Supplier No. 022982.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 555-31-7 | C9H21AlO3 | 204.246 g/mol

Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols. It acts as a precursor to prepare alumina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-31-7
Molecular Formula C9H21AlO3
Molecular Weight (g/mol) 204.246
MDL Number MFCD00008870
InChI Key SMZOGRDCAXLAAR-UHFFFAOYSA-N
Synonym aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
PubChem CID 11143
IUPAC Name aluminum;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Specifications

Melting Point 131°C to 135°C
Density 1.035 g/mL
Boiling Point 135°C (10 mmHg)
Refractive Index 1.0346
Quantity 25 g
UN Number UN3181
Sensitivity Moisture sensitive
Merck Index 14,347
Solubility Information Soluble in ethanol,isopropanol,benzene,toluene,chloroform,carbon tetrachloride and petroleum hydrocarbons.
Formula Weight 204.25
Percent Purity ≥99.99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Aluminum isopropoxide
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum isopropoxide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.