missing translation for 'onlineSavingsMsg'
Learn More

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%

Product Code. 11377328
Change view
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377328 1 kg 1kg
11367328 250 g 250g
11387328 5 x 1 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377328 Supplier Thermo Scientific Alfa Aesar Supplier No. 030715.A1
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32823-06-6

Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32823-06-6
MDL Number MFCD00075314

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 263.9
Concentration or Composition (by Analyte or Components) Al2O3 19.4%, P2O5 78.9%
Density 2.779 g/mL
Chemical Name or Material Aluminum metaphosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.