missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitrate Nonahydrate, Honeywell™

Product Code. 15680820 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15680820 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15680820 Supplier Honeywell Research Chemicals Supplier No. 062751KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7784-27-2
Quantity 1 kg
Molecular Formula AlH18N3O18
Linear Formula Al(NO3)3 · 9H2O
MDL Number MFCD00149132
UN Number NONH for all modes of transport
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
IUPAC Name aluminum;trinitrate;nonahydrate
Molecular Weight (g/mol) 375.129
PubChem CID 24567
Formula Weight 375.13g/mol
Percent Purity ≥98% (KT)
Grade purum p.a.
Chemical Name or Material Aluminum Nitrate Nonahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.