missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitride Rod, Round; Diameter (mm), 10

Product Code. 15471847
Change view
Click to view available options
Quantity:
150 mm
Unit Size:
150mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471847 150 mm 150mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471847 Supplier Thermo Scientific Alfa Aesar Supplier No. 042998.DO
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24304-00-5 | AlN | 40.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24304-00-5
Molecular Formula AlN
Molecular Weight (g/mol) 40.99
MDL Number MFCD00003429
InChI Key PIGFYZPCRLYGLF-UHFFFAOYSA-N
Synonym aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c
PubChem CID 90455
ChEBI CHEBI:50884
IUPAC Name alumanylidyneamine
SMILES N#[Al]

Specifications

Color White to Yellow
Melting Point >2200°C
Appearance White to pale yellow rod
Density 3.26
Quantity 150 mm
Odor Odorless
Form Rod
Chemical Name or Material Aluminum nitride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.