missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum with Over laminated Polyester Transport Signs

Product Code. 11784914 Shop All Brady Products
Change view
Click to view available options
Displayed Text:
Corrosive Substance 8
Environmentally Hazardous Substances
Miscellaneous hazardous substances 9
Organic Peroxide 5.2
Oxidizing substances and organic peroxides 5
Toxic and Harmful Substances 6
Thickness:
0.024 mm
0.271 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Thickness Unit Size
11784914 Corrosive Substance 8 0.271 mm Each
11701003 Environmentally Hazardous Substances 0.271 mm Each
11704924 Miscellaneous hazardous substances 9 0.271 mm Each
11760953 Organic Peroxide 5.2 0.271 mm Each
11710963 Organic Peroxide 5.2 0.024 mm Each
11724914 Oxidizing substances and organic peroxides 5 0.271 mm Each
11744914 Toxic and Harmful Substances 6 0.271 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11784914 Supplier Brady™ Supplier No. 258977
  • ONLINE EXCLUSIVE PRICE
    25.50 €
    24.48 € / Pack of 1
    Save 1.02 €
    4% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number ADR 8-297*297-B7525
Material Aluminum, Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Aluminum with Polyester Overlaminate
For Use With (Application) Pipe markers and safety signs
Height (Metric) 297 mm
Thickness 0.271 mm
Width (Metric) 297 mm
Temperature Range (Metric) -40°C to 140°C
Shape Diamond
Displayed Text Corrosive Substance 8
Product Type Transport Sign
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.