Learn More
Aluminum oxide, alpha-phase, gamma-phase, 99.99% (metals basis)
Description
It is used in stainless steel coating and in microwave applications. It is used as an adsorbent, desiccating agent, grinding or blending agent, abrasive agent and catalyst, and in the manufacture of dental cements and refractories. Industrial applications include electrical and electronic products, metal Products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2045°C |
| Physical Form | Powder |
| Quantity | 100 g |
| Merck Index | 14,356 |
| Solubility Information | Water insoluble. Soluble in hexane,toluene. |
| IUPAC Name | dialuminum;oxygen(2-) |
| Formula Weight | 101.96 |
| Percent Purity | 99.99% |
| Odor | Odorless |
| Assay | (metals basis) |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.