missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, catalyst support, low surface area

Product Code. 11430630
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430630 250 g 250g
11380538 1 kg 1kg
11370538 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430630 Supplier Thermo Scientific Alfa Aesar Supplier No. 043862.30
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Low surface area | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Pellets
Quantity 250 g
IUPAC Name dialuminum;oxygen(2-)
Chemical Name or Material Aluminum Oxide, Catalyst Support

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.