missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, gamma-phase, nanopowder, 99+%

Product Code. 11470001
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470001 100 g 100g
11460001 25 g 25g
11480001 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11470001 Supplier Thermo Scientific Alfa Aesar Supplier No. 044757.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is used in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. It can act as a catalyst in a variety of reactions such as the Claus process. It is used in the production of aluminum, manufacture of abrasives, refractories, ceramics, electrical insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range >99%
Solubility Information Insoluble in water.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Odor Odorless
Density 3.965g/cm3
Chemical Name or Material Aluminum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.