missing translation for 'onlineSavingsMsg'
Learn More

Aluminum perchlorate nonahydrate, Reagent Grade

Product Code. 11391768
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391768 500 g 500g
11381768 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391768 Supplier Thermo Scientific Alfa Aesar Supplier No. 011659.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 81029-06-3 | AlCl3H18O21 | 487.455 g/mol

Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81029-06-3
Molecular Formula AlCl3H18O21
Molecular Weight (g/mol) 487.455
MDL Number MFCD00149130
InChI Key CYRDFMGMNQJULY-UHFFFAOYSA-K
Synonym aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
PubChem CID 16211522
IUPAC Name aluminum;triperchlorate;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]

Specifications

Melting Point 82°C
Physical Form Crystalline
Quantity 500 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 487.46 (325.33 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Aluminum perchlorate nonahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.