Learn More
Aluminum phosphate, Puratronic™, 99.99% (metals basis)
Description
Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >1500°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Appearance | White powder |
| Assay Percent Range | 99.99% (Metal basis) |
| Density | 2.566 |
| Solubility Information | Soluble in concentrated hydrochloric acid,alcohol and nitric acid. Insoluble in water. |
| Formula Weight | 121.95 |
| Chemical Name or Material | Aluminum phosphate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.