missing translation for 'onlineSavingsMsg'
Learn More

Aluminum plate, alloy 6061, 9.53mm (0.375in) thick

Product Code. 15441787
Change view
Click to view available options
Quantity:
30 x 30 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441787 30 x 30 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441787 Supplier Thermo Scientific Alfa Aesar Supplier No. 042128.RF
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

 

Aluminum plate is a raw material for parts as diverse as automotive side panels, storage tank walls, architectural panels and signs. It is also used in manufacturing of fast ferries, yachts, navy or passenger vessels and small pleasure crafts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum plate, Alloy 6061
Name Note 9.53mm (0.375 in.) thick
Quantity 30 x 30 cm
Odor Odorless
Weight ≈2315g/30x30cm
Form Plate
MDL Number MFCD00214039
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.